Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525579

Catalog No. PIPA525579

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

This locus encodes a BTB/POZ domain-containing protein. This domain is known to be involved in protein-protein interactions. Polymorphisms at this locus have been reported to be associated with susceptibility to Restless Legs Syndrome and may also be associated with Tourette Syndrome.
Specifications

Specifications

BTBD9
Polyclonal
PBS with 0.09% sodium azide
Q96Q07, Q8C726
dJ322I12.1, BTB/POZ domain-containing protein 9
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114781, 224671
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
BTBD9
Liquid
BTBD9
KLH conjugated synthetic peptide between 237-262 amino acids from the central region of human BTBD9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit