Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

biotinidase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human BTD protein.

Manufacturer:  Abnova Corporation H00000686B01

Catalog No. 89-004-991


Add to cart

Description

Description

Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency. [provided by RefSeq

Sequence: MAHAHIQGGRRAKSRFVVCIMSGARSKLALFLCGCYVVALGAHTGEESVADHHEAEYYVAAVYEHPSILSLNPLALISRQEALELMNQNLDIYEQQVMTAAQKDVQIIVFPEDGIHGFNFTRTSIYPFLDFMPSPQVVRWNPCLEPHRFNDTEVLQRLSCMAIRGDMFLVANLGTKEPCHSSDPRCPKDGRYQFNTNVVFSNNGTLVDRYRKHNLYFEAAFDVPLKVDLITFDTPFAGRFGIFTCFDILFFDPAIRVLRDYKVKHVVYPTAWMNQLPLLAAIEIQKAFAVAFGINVLAANVHHPVLGMTGSGIHTPLESFWYHDMENPKSHLIIAQVAKNPVGLIGAENATGETDPSHSKFLKILSGDPYCEKDAQEVHCDEATKWNVNAPPTFHSEMMYDNFTLVPVWGKEGYLHVCSNGLCCYLLYERPTLSKELYALGVFDGLHTVHGTYYIQVCALVRCGGLGFDTCGQEITEATGIFEFHLWGNFSTSYIFPLFLTSGMTLEVPDQLGWENDHYFLRKSRLSSGLVTAALYGRLYERD
Specifications

Specifications

biotinidase
Unconjugated
No additive
NM_000060.2
BTD
BTD (NP_000051.1, 1 a.a. ~ 543 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human BTD protein.
BTD
Serum
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
686
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit