Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BTK (p Tyr223), Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178295AF405

Catalog No. NBP178295W


Add to cart

Description

Description

BTK p Tyr223 Polyclonal antibody specifically detects BTK p Tyr223 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

BTK (p Tyr223)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
BTK
A synthetic phosphorylated peptide made to a peptide surrounding amino acid 223 of the human BTK protein [UniProt Q06187]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q06187
Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261
Rabbit
IgG
0.1 ml
Apoptosis, Immunology, Innate Immunity, Phospho Specific
Primary
695
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only