Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BTK (phospho Y223) , Polyclonal, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB51210100UG

Catalog No. NC0145711


Add to cart

Description

Description

Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human. Predicted to work with Mouse
Immunogen: Synthesized phosphopeptide derived from human BTK around the phosphorylation site of tyrosine 223 (A-L-YP-D-Y).

Applications:

ELISA, Western Blotting

Specifications

Specifications

BTK (phospho Y223)
Unconjugated
Rabbit
IgG
Immunology, Signal Transduction
Polyclonal
1mg/mL
Affinity Purified
Synthesized phosphopeptide derived from human BTK around the phosphorylation site of tyrosine 223 (A-L-YP-D-Y).
100μg
Primary
Human, Murine
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit