Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTNL8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524933

Catalog No. PIPA524933


Add to cart

Specifications

Specifications

BTNL8
Polyclonal
PBS with 0.09% sodium azide
Q6UX41
B7-H5 costimulatory molecule, butyrophilin-like protein 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79908
Flow Cytometry, Western Blot
Unconjugated
BTNL8
Liquid
BTNL8
KLH conjugated synthetic peptide between 257-285 amino acids from the central region of human BTNL8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit