Learn More
Bu-1a Mouse anti-Chicken, Unconjugated, Clone: 21-1A4, Southern Biotech™
SDP
Mouse Monoclonal Antibody
Supplier:  Southern Biotechnology Associates 836501
Catalog No. OB836501
Add to Cart
Description
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
Specifications
Bu-1a
Monoclonal
0.5mg/mL
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
Bursa cells from one-day-old H.B15 (Bu-1a/b) strain chickens
RUO
Chicken
Liquid
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
21-1A4
Unconjugated
Mouse
0.5 mg
Primary
Store at 2-8°C
IgG1