Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bub1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525353
Description
BUB1A is a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint.Specifications
Bub1 | |
Polyclonal | |
Unconjugated | |
O43683 | |
BUB1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human BUB1A | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
2 mg/mL | |
BUB1 | |
BUB1A, BUB1L, hBUB1, BUB1 budding uninhibited by benzimidazoles 1 homolog, mitotic checkpoint serine/threonine-protein kinase BUB1, mitotic spindle checkpoint kinase, putative serine/threonine-protein kinase | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
699 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction