Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BXDC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521813

Catalog No. PIPA521813


Add to cart

Description

Description

PA5-21813 targets BXDC1 in WB applications and shows reactivity with Human samples. The PA5-21813 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 217 of BXDC1 (Uniprot IDNo.Q9H7B2). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

BXDC1
Polyclonal
Unconjugated
RPF2
Liquid
RPF2
Recombit fragment corresponding to a region within amino acids 1 and 217 of BXDC1 (Uniprot IDNo.Q9H7B2)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9H7B2
RP11-397G5.1, BXDC1, bA397G5.4, brix domain containing 1, brix domain-containing protein 1, homolog of Rpf2, ribosomal processing factor 2 homolog, ribosome biogenesis protein RPF2 homolog, ribosome production factor 2 homolog
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
84154
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit