Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL6, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132902

Catalog No. PIPA132902

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-32902 detects C 10 from mouse samples.PA1-32902 has been successfully used in Western blot applications.The PA1-32902 immunogen is: Recombinant mouse C-10
Specifications

Specifications

C 10
Polyclonal
PBS with no preservative
P27784
CCL6
Recombinant mouse C-10
Affinity chromatography
RUO
Antibody
Polyclonal
Murine
ELISA, Western Blot
Biotin
Ccl6
Purified
Rabbit
IgG
50μg
Store at -20°C
Primary
20305
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit