Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Cbl Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

$232.00 - $458.00

Specifications

Antigen c-Cbl
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610441
View Documents
BD Biosciences
610441
50μg
Each for $232.00
Add to cart
 
BDB610442
View Documents
BD Biosciences
610442
150μg
Each for $458.00
Add to cart
 
Description

Description

cbl was identified in the genome of a transforming retrovirus from a mouse pre-B cell lymphoma. The product of mammalian c-cbl, p120[cbl], is a widely expressed cytoplasmic protein with several distinctive domains including a RING finger motif and a large proline-rich domain. Although the normal cellular function of c-Cbl has not yet been determined, it associates with many well known signal transduction proteins. In lymphoid cells, c-Cbl is a substrate of TCR-activated protein tyrosine kinases. In addition, stimulation with EGF, PDGF, FGF, or NGF results in tyrosine-phosphorylation of c-Cbl in lymphoid and other cell types. It forms a signaling complex with Grb2 in T cells and is a SH3 and SH2 binding protein which associates with many other cytoplasmic signaling proteins including Btk, Src, fyn, lck, and the β isoform of PI-3 kinase. Thus, c-Cbl may be an important adaptor protein in growth factor and T-cell receptor signaling pathways.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

c-Cbl
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
17
250μg/mL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit