Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB2 Mouse anti-Human, Unlabeled, Clone: 42, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610161

 View more versions of this product

Catalog No. BDB610161


Add to cart

Description

Description

ErbB2 (Neu or Her-2) is a member of the erbB family of growth factor receptors. These factors possess constitutive tyrosine kinase activity and are commonly overexpressed in breast and ovarian carcinomas. While other erbB family members' ligands, such a EGF and NDF, are well characterized, a natural ligand for erbB2 remains unknown. ErbB2 forms heterodimers with erbB1/EGFR, erbB3, and erbB4, and can modulate their ligand affinities. Thus, erbB2 alters the intracellular responses elicited by EGF and NDF. This control is due to the fact that erbB2, when in complex with another erbB family receptor, decelerates the rate of ligand dissociation. Therefore, erbB2 may act as a signaling subunit for other receptors rather than a true growth factor receptor. Due to high sequence homology, this antibody may cross-react with the 180 kDa EGFR.

Specifications

Specifications

ErbB2
42
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Neu, Her-2
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.