Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Jun Mouse anti-Human, Mouse, Rat, Clone: 6E4.4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051076
Description
Anti-c-Jun Antibody, clone 6E4.4 is a Mouse Monoclonal Antibody for detection of c-Jun also known as JNK1 alpha protein kinase, JNK1 beta protein kinase and has been validated in western blot, immunocytochemistry.Specifications
c-Jun | |
Monoclonal | |
Unconjugated | |
NP_002219.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for up to 12 months. | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
6E4.4 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH7.4), 150mM NaCl with 0.05% sodium azide. | |
EC 2.7.11.24 , JNK, JNK-46 , JNK1, JNK1A2, JNK21B1/2, OTTHUMP00000019552, OTTHUMP00000019555, OTTHUMP00000019556, OTTHUMP00000019558, PRKM8, SAPK1 | |
GST-tagged recombinant protein corresponding to c-Jun. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only