Learn More
MilliporeSigma™ anti-c-Kit (961-976), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC34100UG
Catalog No. PC34100UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifically detects c-Kit (961-976) Clone: in Human samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin)
The c-kit proto-oncogene has been recently identified as a member of the receptor-tyrosine kinase family and more specifically a member of the platelet derived growth factor receptor (PDGF-R) family. The c-Kit protein, the normal cellular homolog of the HZ4-feline sarcoma virus v-Kit, encodes a transmembrane receptor with a molecular weight of~145-160kDa. c-Kit has also been shown to be identical with the product of theW locus in mice and as such is integral to the development of mast cells and hematopoiesis. More recently, the ligand of the c-kit receptor (KL) has been identified and characterized and shown to be encoded at the murinesteel (SI) locus.
Specifications
Specifications
c-Kit (961-976)
Unconjugated
purified
a synthetic peptide (GSTASSSQPLLVHDDV) corresponding to amino acids 961-976 of human c-Kit
100μg
−20°C
Polyclonal
Human
Immunofluorescence, Immunohistochemistry (Paraffin)
In PBS.
Rabbit
IgG
RUO
Primary
Recognizes the ∽145kDa c-Kit protein.