Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-c-Myb, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152040

Catalog No. NBP152040


Add to cart

Description

Description

c-Myb Polyclonal antibody specifically detects c-Myb in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

c-Myb
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
MYB
internal region
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize all reported isofoms. (NP_001123645.1; NP_005366.2; NP_001123644.1; NP_001155128.1; NP_001155129.1; NP_001155130.1; NP_001155131.1; NP_001155132.1).
ELISA, Western Blot
Western Blot, Peptide ELISA detection limit 1:8000
NP_001123645.1
c-myb, Cmyb, c-myb protein (140 AA), c-myb_CDS, c-myb10A_CDS, c-myb13A_CDS, c-myb14A_CDS, c-myb8B_CDS, efg, Proto-oncogene c-Myb, transcriptional activator Myb, v-myb avian myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral oncogene homolog (avian)
Goat
IgG
0.1 mg
Cancer
Primary
4602
Bovine, Canine, Human, Mouse, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only