Learn More
NPY Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136143
Catalog No. 01-672-299
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
NPY is a peptide hormone; involved in regulating food intake.
Specifications
Specifications
NPY
Polyclonal
whole serum with 0.1% sodium azide
P01303, P07808
NPY
Synthetic C-PON (residues 68-97).
50 μL
4° C
Primary
24604, 4852
Immunohistochemistry (Frozen)
Unconjugated
NPY
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Rat