Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRP Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen G03011B

Catalog No. PIG03011B


Add to cart

Description

Description

This antibody is an Ig fraction; IgG is > 96.5% of the total protein concentration by SDS-PAGE. Becker titer results showed > 2.65 mg/mL (antigen/antibody).

CRP (C-reactive protein) is a major cyclic, pentameric acute phase protein compound consisting of five identical, noncovalently bound, nonglycosylated subunits. Eeach subunit is made up of 206 amino acids, and has a molecular weight of 24 kDa. CRP is produced by the liver, and its plasma levels rise dramatically during inflammatory processes occurring in the body. CRP is an initiator of classical complement cascade, binds to several nuclear components (chromatin, histones, etc.), plays an important role in innate immunity, and mediates activities associated with pre-immune nonspecific host resistance. Specifically, there is a 100-1000 fold increase of CRP in response to injury, infection or inflammation. CRP is opsonic, an initiator of the classical complement cascade, and an activator of monocytes/macrophages. CRP also binds to several nuclear components including chromatin, histones and snRNP, suggesting that it may play a role as a scavenger during cell necrosis. CRP is involved in several host defense related functions based on its ability to recognize foreign pathogens and damaged cells of the host, and initiates pathogen elimination by interacting with humoral and cellular effector systems in the blood. Patients with elevated basal levels of CRP have been shown to be an increased risk for hypertension and cardiovascular disease.
Specifications

Specifications

CRP
Polyclonal
Unconjugated
CRP
Liquid
CRP
Purified human C-reactive protein
Ion-Exchange Chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunodiffusion, Immunoelectrophoresis, Western Blot
5 mg/mL
PBS with 0.02% sodium azide; pH 7.2
P02741
CRP
Goat
IgG
5 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1401
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit