Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Rel, Polyclonal, Invitrogen™

Catalog No. PIPA130525
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA130525 50 μL
1 options

Catalog No. PIPA130525

Supplier: Thermo Scientific PA130525

Rabbit Polyclonal Antibody

The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA ., RELB (604758), NFKB1 ., and NFKB2 .. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor . (Belguise and Sonenshein, 2007 [PubMed 18037997]).,

PA1-30525 detects c-Rel from human samples.PA1-30525 has been successfully used in immunoprecipitation, Western blot applications.The PA1-30525 immunogen is: c-Rel peptide corresponding to a C-terminus region of human c-Rel
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Rel
Applications ELISA, Gel Shift, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene REL
Gene Accession No. Q04864
Gene Alias C-Rel
Gene Symbols REL
Host Species Rabbit
Immunogen c-Rel peptide corresponding to a C-terminus region of human c-Rel
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.