Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cell division cycle 123 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human C10orf7 protein.

Manufacturer:  Abnova Corporation H00008872B01

Catalog No. 89-006-663


Add to cart

Description

Description

Sequence: MKKEHVLHCQFSAWYPFFRGVTIKSVILPLPQNVKDYLLDDGTLVVSGRDDPPTHSQPDSDDEAEEIQWSDDENTATLTAPEFPEFATKVQEAINSLGGSVFPKLNWSAPRDAYWIAMNSSLKCKTLSDIFLLFKSSDFITRDFTQPFIHCTDDSPDPCIEYELVLRKWCELIPGAEFRCFVKENKLIGISQRDYTQYYDHISKQKEEIRRCIQDFFKKHIQYKFLDEDFVFDIYRDSRGKVWLIDFNPFGEVTDSLLFTWEELISENNLNGDFSEVDAQEQDSPAFRCTNSEVTVQPSPYLSYRLPKDFVDLSTGEDAHKLIDFLKLKRNQQEDD
Specifications

Specifications

cell division cycle 123 homolog (S. cerevisiae)
Unconjugated
No additive
BC001600
C10orf7|D123|FLJ13863
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human C10orf7 protein.
CDC123
Serum
CDC123
C10orf7 (AAH01600.1, 1 a.a. ~ 336 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8872
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit