Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFR1 Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524555
Specifications
SFR1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86XK3 | |
C10orf78, MEI5, MEIR5, bA373N18.1, MEI5 recombination repair protein homolog, SWI5-dependent recombination repair protein 1, meiosis protein 5 homolog, swi5-dependent recombination DNA repair protein 1 homolog | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
119392 |
Flow Cytometry, Western Blot | |
Unconjugated | |
SFR1 | |
Liquid | |
SFR1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human C10orf78 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title