Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
chromosome 10 open reading frame 81, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human C10orf81 protein.
Manufacturer: Abnova Corporation H00079949B01
Description
Sequence: MEQSSPGFRQTHLQDLSEATQDVKEENHYLTPRSVLLELDNIIASSDSGESIETDGPDQVSGRIECHYEPMESYFFKETSHESVDSSKEEPQTLPETQDGDLHLQEQGSGIDWCLSPADVEAQTTNDQKGSASLTVVQLSILINNIPDESQVEKLNVFLSPPDVINYLALTEATGRICVSQWEGPRRLGCIFCHGDHLLAVNDLKPQSLEEVSLFLTRSIQKEKLKLTIGRIPNSETFHAASCMCPSKCQSAAPSQLDKPRLNRAPKRSPAIKKSQQKGARESpecifications
chromosome 10 open reading frame 81 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human C10orf81 protein. | |
C10orf81 | |
FLJ23537/HEL185/bA211N11.2 | |
Mouse | |
50 μL | |
Yes | |
79949 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
BC036365 | |
C10orf81 | |
C10orf81 (AAH36365, 1 a.a. ∼ 282 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title