Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf53 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524404
Description
The specific function of C11orf53 remains unknown.Specifications
C11orf53 | |
Polyclonal | |
C11orf53 | |
uncharacterized protein C11orf53 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
341032 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8IXP5 | |
C11ORF53 | |
KLH conjugated synthetic peptide between 187-215 amino acids from the C-terminal region of human C11orf53 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction