Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf74 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524078

Catalog No. PIPA524078

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The exact function of C11orf74 remains unknown.
Specifications

Specifications

C11orf74
Polyclonal
PBS with 0.09% sodium azide
Q86VG3
HEPIS, uncharacterized protein C11orf74
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
119710
Flow Cytometry, Western Blot
Unconjugated
C11orf74
Liquid
C11ORF74
KLH conjugated synthetic peptide between 190-220 amino acids from the C-terminal region of human C11orf74
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit