Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C12orf30 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523775

Catalog No. PIPA523775


Add to cart

Description

Description

This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
Specifications

Specifications

C12orf30
Polyclonal
PBS with 0.09% sodium azide
Q14CX7, Q8BWZ3
C12orf30, MDM20, NAP1, N-alpha-acetyltransferase 25, NatB auxiliary subunit, N-terminal acetyltransferase B complex subunit NAA25, mitochondrial distribution and morphology 20, natB complex subunit MDM20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231713, 80018
Western Blot
Unconjugated
NAA25
Liquid
NAA25
KLH conjugated synthetic peptide between 720-748 amino acids from the central region of human C12orf30
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit