Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf126 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524053

Catalog No. PIPA524053


Add to cart

Description

Description

Hydrolyzes D-tyrosyl-tRNA(Tyr) into D-tyrosine and free tRNA (Tyr). Could be a defense mechanism against a harmful effect of D-tyrosine (Potential).
Specifications

Specifications

C14orf126
Polyclonal
Unconjugated
DTD2
Liquid
DTD2
KLH conjugated synthetic peptide between 136-164 amino acids from the C-terminal region of human C14orf126
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q96FN9, Q8BHA3
probable D-tyrosyl-tRNA(Tyr) deacylase 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
112487, 328092
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit