Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C15orf29 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526576
Description
The function of the C15orf29 protein has not yet been determined.Specifications
C15orf29 | |
Polyclonal | |
KATNBL1 | |
uncharacterized protein C15orf29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79768 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9H079 | |
KATNBL1 | |
KLH conjugated synthetic peptide between 47-73 amino acids from the N-terminal region of human C15orf29 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction