Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf41 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524193

Catalog No. PIPA524193

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C15orf41
Polyclonal
PBS with 0.09% sodium azide
Q9Y2V0, Q3U4G0
HH114, uncharacterized protein C15orf41
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
399568, 84529
Western Blot
Unconjugated
C15orf41
Liquid
BC052040, C15orf41
KLH conjugated synthetic peptide between 167-193 amino acids from the central region of human C15orf41
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit