Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf59 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524496

Catalog No. PIPA524496


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

Specifications

Specifications

C17orf59
Polyclonal
PBS with 0.09% sodium azide
Q96GS4, Q9D6W8
PRO2472, uncharacterized protein C17orf59
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54785, 71923
Western Blot
Unconjugated
BORCS6
Liquid
BORCS6
KLH conjugated synthetic peptide between 186-215 amino acids from the central region of human C17orf59
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit