Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C19orf18 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526826
Description
Specifications
C19orf18 | |
Polyclonal | |
C19orf18 | |
uncharacterized protein C19orf18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147685 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8NEA5 | |
C19ORF18 | |
KLH conjugated synthetic peptide between 140-168 amino acids from the central region of human C19orf18 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction