Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf21 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524777

Catalog No. PIPA524777


Add to cart

Description

Description

The exact function of C19orf21 remains unknown.
Specifications

Specifications

C19orf21
Polyclonal
PBS with 0.09% sodium azide
Q8IVT2
uncharacterized protein C19orf21
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126353
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
MISP
Liquid
MISP
KLH conjugated synthetic peptide between 63-91 amino acids from the N-terminal region of human C19orf21
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit