Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf26 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526282

Catalog No. PIPA526282


Add to cart

Specifications

Specifications

C19orf26
Polyclonal
PBS with 0.09% sodium azide
Q8N350
DOS, downstream of Stk11, protein Dos
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
255057
Flow Cytometry, Western Blot
Unconjugated
CBARP
Liquid
CBARP
KLH conjugated synthetic peptide between 423-450 amino acids from the central region of human C19orf26
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit