Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C19orf39 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525460
Description
Specifications
C19orf39 | |
Polyclonal | |
SWSAP1 | |
C19orf39, SWS1AP1, ZSWIM7AP1, ATPase SWSAP1, SWIM-type zinc finger 7-associated protein 1, SWS1-associated protein 1, zinc finger, SWIM-type containing 7 associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126074, 66962 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q6NVH7, Q8VCI7 | |
SWSAP1 | |
KLH conjugated synthetic peptide between 84-110 amino acids from the central region of human C19orf39 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction