Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KXD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526148

Catalog No. PIPA526148

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

The function of the C19orf50 protein has not yet been determined.
Specifications

Specifications

KXD1
Polyclonal
PBS with 0.09% sodium azide
Q9BQD3
C19orf50, KXDL, MST096, MSTP096, UPF0459 protein C19orf50
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79036
Immunohistochemistry, Western Blot
Unconjugated
KXD1
Liquid
KXD1
KLH conjugated synthetic peptide between 59-85 amino acids from the central region of human C19orf50
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit