Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
chromosome 1 open reading frame 57 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human C1orf57 protein.
Manufacturer: Abnova Corporation H00084284B01
Description
Sequence: MARHVFLTGPPGVGKTTLIHKASEVLKSSGVPVDGFYTEEVRQGGRRIGFDVVTLSGTRGPLSRVGLEPPPGKRECRVGQYVVDLTSFEQLALPVLRNADCSSGPGQRVCVIDEIGKMELFSQLFIQAVRQTLSTPGTIILGTIPVPKGKPLALVEEIRNRKDVKVFNVTKENRNHLLPDIVTCVQSSRKSpecifications
chromosome 1 open reading frame 57 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human C1orf57 protein. | |
C1orf57 | |
FLJ11383/MGC13186/RP4-678E16.2 | |
Mouse | |
50 μL | |
Kinases | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
No additive | |
NM_032324.1 | |
C1orf57 | |
C1orf57 (NP_115700.1, 1 a.a. ∼ 190 a.a) full-length human protein. | |
RUO | |
Yes | |
84284 | |
Serum |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title