Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

integrator complex subunit 7, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human C1orf73 protein.

Manufacturer:  Abnova Corporation H00025896B01

Catalog No. 89-007-353


Add to cart

Description

Description

INTS7 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM

Sequence: MASNSTKSFLADAGYGEQELDANSALMELDKGLRSGKLGEQCEAVVRFPRLFQKYPFPILINSAFLKLADVFRVGNNFLRLCVLKVTQQSEKHLEKILNVDEFVKRIFSVIHSNDPVARAITLRMLGSLASIIPERKNAHHSIRQSLDSHDNVEVEAAVFAAANFSAQSKDFAVGICNKISEMIQGLATPVDLKLKLIPILQHMHHDAILASSARQLLQQLVTSYPSTKMVIVSLHTFTLLAASSLVDTPKQIQLLLQYLKNDPRKAVKRLAIQDLKLLANKTPHTWSRENIQALCECALQTPYDSLKLGMLSVLSTLSGTIAIKHYFSIVPGNVSSSPRSSDLVKLAQECCYHNNRGIAAHGVRVLTNITVSCQEKDLLALEQDAVFGLESLLVLCSQDDSPGAQATLKIALNCMVKLAKGRPHLSQSVVETLLTQLHSAQDAARILMCHCLAAIAMQLPVLGDGMLGDLMELYKVIGRSATDKQQELLVSLATVIFVASQKALSVESKAVIKQQLESVSNGWTVYRIARQASRMGNHDMAKELYQSLLTQVASEHFYFWLNSLKEFSHAEQCLTGLQEENYSSALSCIAESLKFYHKGIASLTAASTPLNPLSFQCEFVKLRIDLLQAFSQLICTCNSLKTSPPPAIATTIAMTLGNDLQRCGRISNQMKQSMEEFRSLASRYGDLYQASFDADSATLRNVELQQQSCLLISHAIEALILDPESASFQEYGSTGTAHADSEYERRMMSVYNHVLEEVESLNRKYTPVSYMHTACLCNAIIALLKVPLSFQRYFFQKLQSTSIKLALSPSPRNPAEPIAVQNNQQLALKVEGVVQHGSKPGLFRKIQSVCLNVSSTLQSKSGQDYKIPIDNMTNEMEQRVEPHNDYFSTQFLLNFAILGTHNITVESSVKDANGIVWKTGPRTTIFVKSLEDPYSQQIRLQQQQAQQPLQQQQQRNAYTRF
Specifications

Specifications

integrator complex subunit 7
Unconjugated
No additive
NM_015434.1
C1orf73|DKFZp434B168|INT7
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
25896
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human C1orf73 protein.
INTS7
Serum
INTS7
C1orf73 (NP_056249.1, 1 a.a. ~ 962 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit