Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1q Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA129720
Description
For tissue staining, antigen retrieval is recommended using Proteinase K for 30 min at 37oC.
C1q Polyclonal antibody specifically detects C1q in Hamster, Human samples. It is validated for Functional Assay, Immunohistochemistry (Paraffin).Specifications
C1q | |
Polyclonal | |
whole serum with 0.05% sodium azide | |
P02745, P02746, P02747 | |
C1QA, C1QB, C1qc | |
C1q isolated from the normal human serum. | |
1 mL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG |
Functional Assay, Immunohistochemistry (Paraffin) | |
FITC | |
C1qc | |
AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B | |
Rabbit | |
Ig fraction | |
RUO | |
100766409, 100766700, 100769576, 712, 713, 714 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction