Learn More
C1q Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA514208
Catalog No. PIPA514208
Add to cart
Description
Description
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
Specifications
Specifications
C1q
Polyclonal
Unconjugated
C1QA
Liquid
C1QA
KLH conjugated synthetic peptide between 198-227 amino acids from the C-terminal region of human C1QA
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
P02745
Complement C1q subcomponent subunit A; C1QA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
712