Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QTNF5, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human C1QTNF5 protein.

Manufacturer:  Abnova Corporation H00114902B01P

Catalog No. 89-002-865


Add to cart

Description

Description

A bicistronic transcript encodes the products of the membrane frizzled-related gene and the complement C1q tumor necrosis factor-related gene 5, located on chromosome 11

Sequence: MRPLLVLLLLGLAAGSPPLDDNKIPSLCPGHPGLPGTPGHHGSRGLPGRDGRDGRDGAPGAPGEKGEGGRPGLPGPRGDPGPRGEAGPAGPTGPAGECSVPPRSAFSAKRSESRVPPPSDAPLPFDRVLVNEQGHYDAVTGKFTCQVPGVYYFAVHATVYRASLQFDLVKNGESIASFFQFFGGWPKPASLSGGAMVRLEPEDQVWVQVGVGDYIGIYASIKTDSTFSGFLVYSDWHSSPVFA
Specifications

Specifications

C1QTNF5
Unconjugated
In 1X PBS, pH 7.4
BC029485.1
CTRP5|DKFZp586B0621|LORD
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human C1QTNF5 protein.
C1QTNF5
Purified
C1QTNF5
C1QTNF5 (AAH29485.1, 1 a.a. ~ 243 a.a) full-length human protein.
RUO
Primary
114902
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit