Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1r Mouse anti-Human, Clone: 269104, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523969

Catalog No. PIMA523969


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human (rh) C1s or rhC1q R1 is observed. In Western blots, ths antibody recognizes rhC1r and the A/heavy chain. Reconstitute at 0.5 mg/mL in sterile PBS.

C1r B chain is a serine protease that combines with C1q and C1s to form C1, the first component of the classical pathway of the complement system.
Specifications

Specifications

C1r
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P00736
complement C1r subcomponent
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
715
Immunoprecipitation, Western Blot
269104
Unconjugated
C1R
Lyophilized
C1R
Mouse myeloma cell line NS0-derived recombit human Complement Component C1r Ser18-Asp705
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit