Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromosome 2 open reading frame 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human C2orf3 protein.

Manufacturer:  Abnova Corporation H00006936B01

Catalog No. 89-006-332


Add to cart

Description

Description

The first mRNA transcript isolated for this gene was part of an artificial chimera derived from two distinct gene transcripts and a primer used in the cloning process (see Genbank accession M29204). A positively charged amino terminus present only in the chimera was determined to bind GC-rich DNA, thus mistakenly thought to identify a transcription factor gene. [provided by RefSeq

Sequence: MAHRPKRTFRQRAADSSDSDGAEESPAEPGAPRELPVPGSAEEEPPSGGGRAQVAGLPHRIRGPRGRGRVWASSRRATKAAPRADEGSESRTLDVSTDEEDKIHHSSESKDDQGLSSDSSSSLGEKELSSTVKIPDAAFIQAARRKRELARAQDDYISLDVQHTSSISGMKRESEDDPESEPDDHEKRIPFTLRPQTLRQRMAEESSMDLPIYED
Specifications

Specifications

chromosome 2 open reading frame 3
Unconjugated
No additive
BC000853
DNABF|GCF|TCF9
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6936
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human C2orf3 protein.
C2orf3
Serum
C2orf3
C2orf3 (AAH00853, 1 a.a. ~ 215 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit