Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf49 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525113

Catalog No. PIPA525113

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

C2orf49
Polyclonal
PBS with 0.09% sodium azide
Q9BVC5
asw, ashwin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79074
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
C2orf49
Liquid
C2ORF49
KLH conjugated synthetic peptide between 62-90 amino acids from the N-terminal region of human C2orf49
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit