Learn More
anti-C2orf74, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232313
Catalog No. NBP232313
Add to cart
Description
Description
C2orf74 Polyclonal antibody specifically detects C2orf74 in Human samples. It is validated for Western Blot.
Specifications
Specifications
C2orf74
0.47 mg/mL
Western Blot 1:1000
A8MZ97
uncharacterized protein C2orf74, chromosome 2 open reading frame 74
Rabbit
IgG
Protein A purified
RUO
Primary
339804
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
C2orf74
This C2orf74 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 164-194 amino acids from the C-terminal region of human C2orf74.
21.915 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human