Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128064

Catalog No. PA128064

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28064 detects C3 from mouse samples.PA1-28064 has been successfully used in immunohistochemistry (frozen tissue) and Western blot applications.The PA1-28064 immunogen is a proprietary sequence.
Specifications

Specifications

C3
Polyclonal
Unconjugated
C3
Purified
C3
proprietary sequence
Protein G
RUO
Antibody
Polyclonal
Murine
Immunohistochemistry (Frozen), Western Blot
0.1mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P01027
AI255234, ASP, Plp
Rabbit
IgG
50μg
Store at 4°C
Primary
12266
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit