Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3a Receptor Mouse anti-Human, Clone: 534625, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524235

Catalog No. PIMA524235


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

This polyclonal antibody was raised in Rabbit against C3AR / C3a Receptor. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human C3a Receptor. BLAST analysis of the peptide immunogen showed no homology with other human proteins, except ADRA1A (40%).
Specifications

Specifications

C3a Receptor
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q16581
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Mouse
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
719
Flow Cytometry
534625
Unconjugated
C3AR1
Lyophilized
C3AR1
NS0 mouse myeloma cell line transfected with human C3a R Met1-Val482
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit