Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf31 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525310

Catalog No. PIPA525310

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C3orf31
Polyclonal
PBS with 0.09% sodium azide
Q96BW9, Q3TUH1
C3orf31, TAM41, MMP37-like protein, mitochondrial, mitochondrial translocator assembly and maintece protein 41 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
132001, 68971
Flow Cytometry, Western Blot
Unconjugated
TAMM41
Liquid
TAMM41
KLH conjugated synthetic peptide between 180-208 amino acids from the central region of human C3orf31
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit