Learn More
C3orf75 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525087
Catalog No. PIPA525087
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The function of this protein has not been specifically defined.
Specifications
Specifications
C3orf75
Polyclonal
Unconjugated
ELP6
Liquid
ELP6
KLH conjugated synthetic peptide between 223-252 amino acids from the C-terminal region of human C3orf75
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q0PNE2
TMEM103, UPF0405 protein C3orf75, angiotonin-transactivated protein 1, transmembrane protein 103
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54859