Learn More
anti-C5L2/GPR77, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131240
Catalog No. NBP131240
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
C5L2/GPR77
1.0mg/mL
Western Blot 1:500-1:3000
Q9P296
C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (86%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
C5AR2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human C5L2. The exact sequence is proprietary.
36 kDa
100 ul
GPCR
Primary
27202
Human