Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-C5L2/GPR77, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP131489

Catalog No. NBP131489

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C5L2/GPR77
1.0mg/mL
Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Q9P296
C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (83%). Not yet tested in any other species.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0) and 50% Glycerol with 0.01% Thimerosal
Affinity Purified
C5AR2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human C5L2(GPR77). The exact sequence is proprietary.
36 kDa
100 ul
GPCR
Primary
27202
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only