Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf28A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525368

Catalog No. PIPA525368

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

CG028 belongs to the UPF0550 family.
Specifications

Specifications

C7orf28A
Polyclonal
PBS with 0.09% sodium azide
P86791, Q8C1Y8
C7orf28A, CCZ1A, H_DJ1163J12.2, H_NH0577018.2, vacuolar fusion protein CCZ1 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231874, 51622
Immunohistochemistry, Western Blot
Unconjugated
CCZ1
Liquid
CCZ1
KLH conjugated synthetic peptide between 239-265 amino acids from the central region of human C7orf28A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit