Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R35 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525117

Catalog No. PIPA525117

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The function of this protein has not been specifically defined.
Specifications

Specifications

PPP1R35
Polyclonal
Unconjugated
PPP1R35
Liquid
PPP1R35
KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human C7orf47
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8TAP8, Q9D8C8
C7orf47, UPF0683 protein C7orf47, protein phosphatase 1 regulatory subunit 35
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
221908, 69871
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit