Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromosome 9 open reading frame 89, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human C9orf89 protein.

Manufacturer:  Abnova Corporation H00084270B01

Catalog No. 89-008-325


Add to cart

Description

Description

Sequence: MTDQTYCDRLVQDTPFLTGHGRLSEQQVDRIILQLNRYYPQILTNKEAEKFRNPKASLRVRLCDLLSHLQRSGERDCQEFYRALYIHAQPLHSRLPSRHALQNSDCTELDSGSQSGELSNRGPMSFLAGLGLAVGLALLLYCYPPDPKGLPGTRRVLGFSPVIIDRHVSRYLLAFLADDLGGL
Specifications

Specifications

chromosome 9 open reading frame 89
Unconjugated
No additive
NM_032310.3
BinCARD|MGC110898|MGC11115|bA370F5.1
Mouse
50 uL
Kinases
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human C9orf89 protein.
C9orf89
Serum
C9orf89
C9orf89 (NP_115686.3, 1 a.a. ~ 183 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
84270
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit